CLS4423 Sigma

Corning® 96 well PP 1.2 mL cluster tubes

96 well polypropylene cluster tubes, individual tube format, non-treated, sterile, clear, 96 tubes/rack, 4800/cs

DOWNLOAD MSDS (PDF)

Synonym: 96 multiwell plates, 96 well microplates, 96 well plates, Corning tubes, cap tubes, polypropylene tubes

Properties

Related Categories Cell Culture Supplies, Culture Tubes, Plastic, Labware More...
material   polypropylene
  clear round bottom wells
sterility   sterile
feature   lid: No
  Plate format: 96 well cluster tubes
packaging   case of 4800
  pack of 960
mfr. no.   Corning, 4423
tube capacity   1.2 mL
tube working volume   ≤1mL
color   clear plate
  clear
matrix active group   non-treated surface treatment

Description

Legal Information

Corning is a registered trademark of Corning, Inc.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?