Popular Documents: Bulletin (PDF)
| Related Categories | Cell Biology, Cell Signaling and Neuroscience, Cytoskeleton and Extracellular Matrix, Kits |
| usage | sufficient for 100 tests |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... B4GALT1(2683) |
The kit provides a convenient method for quantitative estimation of the UDP-galactosyltransferase activitity in cell or tissue extracts.
The kit assay is based on the transfer of radioactively labeled galactose from UDP-galactose to N-acetyl-D-glucosamine by UDP-galactosyltransferase
UDP-galactosyltransferase (β4GalT1) is a trans-Golgi membrane bound protein that participates in protein processing in the Golgi apparatus. UDP-galactosyl-transferas
|
Description |
Product # |
Add to Cart |
|---|---|---|
| Assay Buffer 500 μL | ||
| Manganese Chloride Solution 500 μL | ||
| Acceptor Solution 500 μL | ||
| UDP-Gal Solution 500 μL | ||
| Cleanup Resin 50 mL | ||
| Mini-Columns, capacity 5 mL, overall H 1.5 in. 100 each | ||
| Elution Solution 100 mL |
Customers Also Viewed
≥0.6 U/mg
powder, slightly yellow, ≥5 U/g
lyophilized powder
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?