Synonym: Anti-H3K4me2
MDL number MFCD06798436
Popular Documents: Datasheet (PDF)
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Epigenetics, Antibodies for Gene Regulation, |
| species reactivity | Caenorhabditis elegans, Arabidopsis thaliana, human, Drosophila, bovine, frog, rat, chicken, mouse |
| application(s) | ChIP: suitable |
| microarray: suitable | |
| western blot: 1:1,000-1:2,000 using whole extract of human epitheloid carcinoma HeLa cell line and mouse fibroblast NIH3T3 cell line | |
| clone | polyclonal |
| antibody form | IgG fraction of antiserum |
| form | buffered aqueous solution |
| mol wt | antigen mol wt 17 kDa |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information |
human ... H3F3A(3020), H3F3B(3021), HIST1H3A(8350), HIST1H3B(8358), HIST1H3C(8352), HIST1H3D(8351), HIST1H3E(8353), HIST1H3F(8968), HIST1H3G(8355), HIST1H3H(8357), HIST1H3I(8354), HIST1H3J(8356), HIST2H3A(333932), HIST2H3C(126961), HIST3H3(8290) mouse ... H3f3a(15078), H3f3b(15081), Hist1h3a(360198), Hist1h3b(319150), Hist1h3c(319148), Hist1h3d(319149), Hist1h3e(319151), Hist1h3f(260423), Hist1h3g(97908), Hist1h3h(319152), Hist1h3i(319153), Hist2h3b(319154), Hist2h3c1(15077), Hist2h3c2(97114) rat ... H3f3b(117056) |
synthetic methylated peptide corresponding to amino acids 1-8 [diMe-Lys4] of human histone H3, conjugated to KLH. This sequence is identical in many species including mouse, rat, bovine, chicken, frog, Drosophila, C. elegans, tetrahymena, and Arabidopsis thaliana histone H3.
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
ChIP validated
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?