Synonym: Anti-Kiaa0478, Anti-Zinc finger and btb domain containing 40, Anti-Znf923
Human Protein Atlas Number HPA042386 Human Protein Atlas characterization data
| Related Categories | Alphabetical Index, Antibodies, Prestige Antibodies, Prestige Polyclonal Antibodies, Primary Antibodies, |
| species reactivity | human |
| application(s) | immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable |
| protein array: suitable | |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | buffered aqueous glycerol solution |
| grade | Prestige Antibodies® Powered by Atlas Antibodies |
| immunogen sequence |
ATLETILRHNQLILEAIQQKIEYKL |
| shipped in | wet ice |
| storage temp. | −20°C |
| Gene Information | human ... ZBTB40(9923) |
zinc finger and BTB domain containing 40 recombinant protein epitope signature tag (PrEST)
Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.
Anti-ZBTB40 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project (www.proteinatlas.org)
. Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using protein array and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Prestige Antibodies is a registered trademark of Sigma-Aldrich Co. LLC
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?