Synonym: Triacylglycerol acylhydrolase, Triacylglycerol lipase
CAS Number 9001-62-1
EC Number 232-619-9
MDL number MFCD00131509
Popular Documents: Specification Sheet (PDF) | RAMAN FTIR (PDF)
| Related Categories | 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Diagnostic and Analytical Enzymes, |
| type | Type XII |
| form | lyophilized powder |
| composition | Protein, 50-75% Lowry |
| storage temp. | −20°C |
Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).
One unit will hydrolyze 1.0 microequivalent of fatty acid from a triglyceride in 1 hr at pH 7.7 at 37 °C using olive oil (30 minute incubation).
Customers Also Viewed
lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Type VI-S, ≥20,000 units/mg protein, lyophilized powder
≥20,000 U/g
solution, ≥100,000 U/g
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?