Synonym: HGFR, RCCP2
Popular Documents: Datasheet (PDF) | Specification Sheet (PDF)
| Related Categories | Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, |
| recombinant | expressed in baculovirus infected Sf9 cells |
| grade | PRECISIO® Kinase |
| assay | ≥70% (SDS-PAGE) |
| form | buffered aqueous glycerol solution |
| activity | 37-55 nmol/min·mg |
| mol wt | mol wt ~81 kDa |
| shipped in | dry ice |
MET is a proto-oncogene that encodes a transmembrane growth factor receptor which is a heterodimer of two disulfide linked chains of 50 kDa (alpha) and 145 kDa (beta). MET is widely expressed in the kidney, brain, lung, skin, and embryonic tissue. Hepatocyte growth factor (HGF) binds to MET and activates its tyrosine kinase activity. MET is overexpressed and activated in a variety of human cancers including pancreatic, colon, gastric, cervical and ovariancancers and has been shown to be involved in tumor cell migration and invasion.
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?