M5561 Sigma

Greiner high and medium binding 96 well plates

clear polystyrene washer flat bottom (with micro-clear bottom), surface treatment medium binding, non-sterile

DOWNLOAD MSDS (PDF)

Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Properties

Related Categories 96 Well Plates, Greiner, Greiner 96-Well Multiwell Plates, Greiner High & Medium Binding 96-Well Plates, Greiner Multiwell Plates,
material   clear polystyrene washer flat bottom (with micro-clear bottom)
  white polystyrene plate
sterility   non-sterile
mfr. no.   Greiner 655095
packaging   case of 40
maximum volume   392 μL
size   96 wells
working volume   340 μL
color   white plate
matrix active group   medium binding surface treatment

Description

General description

LUMITRAC 200 96W Microplate, well working volume 25-340 μL; total well volume 392 μL.

Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Other Notes

96 well flat (F) solid bottom; medium binding microplate, chimney style, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Suitability

White plates minimize cross talk in luminescence procedures. LUMITRAC 200 will bind up to 200 ng/cm2. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

LUMITRAC is a trademark of Greiner Bio-One GmbH

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?