M8698 Sigma

Methionine Sulfoxide Reductase A, histidine-tagged human

recombinant, expressed in E. coli, ≥8 units/mg protein, buffered aqueous solution

DOWNLOAD MSDS (PDF)

Synonym: Methionine S-oxide reductase (S-form oxidizing), MsrA, peptide-Methionine sulfoxide reductase, protein-Methionine-S-oxide-reductase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cell Stress, Enzymes, Nitric Oxide and Cell Stress,
recombinant   expressed in E. coli
assay   ≥90% (SDS-PAGE)
form   buffered aqueous solution
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... MSRA(4482)

Description

Analysis Note

The assay is based on the ability of MsrA to reduce methionine sulfoxide, which is measured by monitoring the level of NADPH oxidation in the presence of a thioredoxin-regenerating system.

Unit Definition

One unit will cause the oxidation of 1 μmole of NADPH per minute at 37 °C at pH 7.6.

Physical form

Supplied in a solution of 50 mM Tris, pH 7.6, and 150 mM NaCl.

Biochem/physiol Actions

Methionine (Met) residues of proteins are readily oxidized to methionine sulfoxide (MetO), especially under oxidative stress conditions. Oxidative alteration of Met to R/S-Met(O) sterioisomers is reversed by methionine sulfoxide reductases: MsrA reduces S-MetO and MsrB reduces R-MetO, which may prevent irreversible oxidative protein damage and thus extend an organism′s life span.1,2 A series of proteins have been identified as substrates of MsrA, these include Calmodulin and HIV protease. There is evidence showing a connection between MsrA and Alzheimer′s disease.3

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?