N3258 Sigma

Nicotinamide N-oxide

≥98%

DOWNLOAD MSDS (PDF)

Synonym: 3-Pyridinecarboxamide, Oxynicotinamide

  • CAS Number 1986-81-8

  • Empirical Formula (Hill Notation) C6H6N2O2

  • Molecular Weight 138.12

  •  EC Number 217-859-4

  •  MDL number MFCD00006202

  •  PubChem Substance ID 24897634

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories Biochemicals and Reagents, NAD(P)(H) Analogs, NAD(P)(H) and Analogs, Nucleosides, Nucleotides, Oligonucleotides
assay   ≥98%
mp   291-293 °C (dec.)(lit.)

Description

Biochem/physiol Actions

Nicotinamide N-oxide is a niacin-related compound used in studies of mechanisms of niacin-related granulocyte differentiation of cells such as human promyelocytic leukemia cells (HL-60). Nicotinamide N-oxide reduced expression of c-myc in HL-60 cell line.

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. N3258.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Price and Availability

Customers Also Viewed

Nicotinamide

≥99.5% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?