NBAA5 Sigma

Panorama® Antibody Array-Neurobiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories Antibody Arrays, Cell Biology, Neurobiology Array, Panorama Antibody Array- Neurobiology, Protein Arrays More...
species reactivity   human, mouse, rat
shipped in   wet ice
storage temp.   2-8°C

Description

Components

224 antibodies spotted in 32 sub-arrays, each containing duplicate spots of the antibodies, as well as duplicate positive control spots (a monoclonal antibody that recognizes Cy3 and Cy5) and several negative controls. Antibodies are spotted in high density to ensure strong signals. Antibodies are also available individually from Sigma for more extensive analysis of your sample. Refer to Antibody List for specific catalog number(s). sigma.com/nbaa5

Compatibility

Cell or tissue extracts, blood plasma or serum

Other Notes

Please note that featured antibody content is lot specific and is subject to change.

For more information see Protein Array Literature References

Physical form

Nitrocellulose glass slide, proprietary treatment to minimize background staining.

Application

The array can be used for comparing protein expression profiles between two samples (test vs. reference samples). Each sample is labeled with a different dye (such as Cy3 or Cy5) and the 2 samples are applied simultaneously on the array, The expression profile of each sample is recorded individually and compared.

General description

The Panorama Antibody Microarray - Neurobiology is a proteomics tool to measure the relative level of protein expression in cell or tissue extracts. The array contains 224 different antibodies on nitrocellulose coated glass slides. These antibodies represent targets to highly studied neurodegenerative diseases, such as Alzheimer′s, Huntington′s, and Parkinson′s. Also represented on the array are targets involved in neuron development.

Legal Information

Panorama is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Kit component also available separately

Description

Product #

Add to Cart

Extraction/Labeling Buffer E0655
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity) P5726
Phosphate buffered saline, pH 7.4, contains TWEEN® 20, dry powder P3563

Kit component only

Description

Product #

Add to Cart

Panorama Antibody Slide- Neurobiology    
QuadiPERM Cell Culture Vessel 2EA    
Protease Inhibitor Cocktail 1VL    
Benzonase®, Ultrapure 1000UN    
Array Incubation Buffer 20ML    
SigmaSpin Post-Reaction Clean-Up Columns 8EA    
Microcentrifuge collection tubes 16EA    
Panorama Antibody List- Neurobiology 1EA    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?