Synonym: Ampholytes
CAS Number 70852-56-1
MDL number MFCD00145874
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Capillary Electrophoresis, Capillary Isoelectric Focusing, Carrier Ampholytes, |
| pH range | 8.0 - 10.5 |
| capacity | 0.35 mmol/pH·mL buffering capacity |
| storage temp. | 2-8°C |
IEF carrier ampholytes
Pharmalyte is a registered trademark of GE Healthcare
Customers Also Viewed
pH range 3.0 - 10.0
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?