CAS Number 93908-02-2
Empirical Formula (Hill Notation) C27H21Cl2N3O7
Molecular Weight 570.38
MDL number MFCD01718312
PubChem Substance ID 24899375
Popular Documents: Product Information Sheet (PDF)
| Related Categories | Antibiotics, Antibiotics A to Z, Antibiotics N-S, Antibiotics by Application, Antifungal, |
| biological source | from Saccharothrix aerocolonigenes |
| assay | ≥98% (HPLC) |
| form | solid |
| storage condition | protect from light |
| color | yellow |
| solubility | DMSO: ≥3 mg/mL |
| THF: soluble |
Rebeccamycin is an antibiotic composed of a halogenated indolocarbazole chromophore linked via N-glycosidic bond to a glucose derivative. As an antitumor agent,1 it is cytotoxic to tumor cell lines such as human lung adenocarcinoma, colon carcinoma and nasopharyngeal carcinoma, etc.2 Rebeccamycin intercalates into the DNA and is an inhibitor of topoisomerase I. It stabilizes the covalent topoisomerase I-DNA intermediate (the cleavable complex) resulting in increased DNA strand breaks.3
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?