S2151 Sigma
Spill trays
L × W 21 in. (53.5 cm) × 44 in. (112 cm)
DOWNLOAD MSDS (PDF)
Properties
Related Categories Labware, Safety Equipment, Spill Tray More...
material   white PVC (rigid)
L × W   21 in. (53.5 cm) × 44 in. (112 cm)
Description
General description
In sizes for desktop or hood use. Rounded corners are easily cleaned. Tapered outer edge provides comfortable wrist rest and stabilizes tray against movement and tipping. Seamless construction provides spill containment for most common laboratory chemicals.
When used with disposable fitted liners (below) and specially designed workshields (q.v.), these units provide a valuable additional safety margin for handling radioactive or biohazardous materials.
Price and Availability
Customers Also Viewed
Plastic development tray
size 8 1/4 in. × 7 in. × 1 3/8 in.
pkg of 1 ea
Spill trays
L × W 21 in. (53.5 cm) × 27 in. (68.5 cm)
Techware PP bench tray
pkg of 1 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?