| Related Categories | 2C-2E, Alphabetical Index, Antibodies, Clone Index, Primary Antibodies, |
| species reactivity | human |
| application(s) | capture ELISA: suitable |
| immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable | |
| indirect ELISA: suitable | |
| western blot: suitable | |
| clone | 2D5, monoclonal |
| concentration | ~0.44 mg/mL |
| antibody form | purified immunoglobulin |
| form | buffered aqueous solution |
| isotype | IgG1κ |
| immunogen sequence |
MGCIKSKENKSPAIKYRPENTPEPV |
| mol wt | antigen mol wt ~85.84 kDa |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... YES1(7525) |
| NCBI accession no. |
BC048960 |
YES1 (AAH48960, 1 a.a. ~ 544 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Mouse monoclonal antibody raised against a full length recombinant YES1.
Clear, colorless solution in phosphate buffered saline, pH 7.2
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?