Synonym: Anti-CSNB1B, Anti-DKFZp686H1993, Anti-GPRC1F, Anti-Glutamate receptor, metabotropic 6, Anti-MGLUR6
MDL number MFCD03097014
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Neurotransmission, Antibodies to Glutamate Receptors, |
| species reactivity | chicken, rabbit, canine, human, mouse, rat |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | lyophilized powder |
| immunogen sequence |
AGGLTLGGLFPVHARGAAGRACGQL |
| storage temp. | −20°C |
| Gene Information | human ... GRM6(2916) |
Peptide region of the protein sequence according to NP_000834.
Lyophilized from PBS buffer with 2% sucrose
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Customers Also Viewed

affinity isolated antibody, buffered aqueous solution

~1 mg/mL, affinity isolated antibody, lyophilized powder
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?