| Related Categories | AS-ATG, Alphabetical Index, Antibodies, Primary Antibodies |
| species reactivity | rat, human |
| application(s) | indirect ELISA: suitable |
| western blot: suitable | |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | lyophilized powder |
| storage temp. | −20°C |
| Gene Information | human ... Atf2(81647) |
| NCBI accession no. |
NM_031018 |
Antibody is lyophilized from PBS buffer with 2% sucrose.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?