Synonym:
Anti-
| Related Categories | Alphabetical Index, Antibodies, Antibodies to Human Plasma Proteins, LRR-LZ, Primary Antibodies, |
| species reactivity | human |
| application(s) | indirect ELISA: suitable |
| western blot: suitable | |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | buffered aqueous solution |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... LTF(4057) |
Peptide with sequence C-ENYKSQQSSDPDP from the internal region of the protein sequence according to NP_002334.2.
Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?