SAB3300039 Sigma

Monoclonal Anti-Pdx1 antibody produced in mouse

clone 5D5, purified immunoglobulin, buffered aqueous glycerol solution

DOWNLOAD MSDS (PDF)

Synonym: Anti-GSF, Anti-IDX-1, Anti-IDX-2, Anti-IPF1, Anti-IUF1, Anti-IUF2

Properties

Related Categories 5, Alphabetical Index, Antibodies, Antibodies against Proteins, Bioactives, Markers, and Receptors for Stem Cell Biology, Antibodies to Drug Transport Proteins,
species reactivity   human
application(s)   western blot: suitable
clone   5D5, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous glycerol solution
isotype   IgG1
mol wt   antigen mol wt 30.6 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... PDX1(3651)
NCBI accession no.   NM_000209

Description

Immunogen

Recombinant protein expressed in E. coli corresponding to amino acids 1-283 of human Pdx1

Antigen Background

Pdx1 is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. It is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in PDX1 gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).

Physical form

Solution in phosphate buffered saline containing 1% bovine serum albumin, 50% glycerol, and 0.02% sodium azide.

Price and Availability

Anti-Mouse 2° Antibody Best Sellers


Prod. No.ConjugateSecondary Antibody Description
A4416HRPAnti-Mouse IgG (whole molecule)-Peroxidase
A3562APAnti-Mouse IgG (whole molecule)-Alkaline Phosphatase
F0257FITCAnti-Mouse IgG (whole molecule)-FITC
B7264BiotinAnti-Mouse IgG (whole molecule)-Biotin





Customers Also Viewed

Monoclonal Anti-Pdx1 antibody produced in mouse

clone 2A12, purified immunoglobulin, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?