SAB4100016 Sigma

Monoclonal Anti-C15orf40 antibody produced in mouse

culture supernatant

DOWNLOAD MSDS (PDF)

Synonym: Anti-Chromosome 15 open reading frame 40, Anti-MGC29937

Properties

Related Categories Alphabetical Index, Antibodies, C, C1-C19, Clone Index,
species reactivity   human
application(s)   western blot: suitable
clone   C15orf40-B23, monoclonal
antibody form   culture supernatant
form   aqueous solution
isotype   IgG1
mol wt   antigen mol wt 16.2 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... C15orf40(123207)

Description

Immunogen

C15orf40 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Price and Availability

Anti-Mouse 2° Antibody Best Sellers


Prod. No.ConjugateSecondary Antibody Description
A4416HRPAnti-Mouse IgG (whole molecule)-Peroxidase
A3562APAnti-Mouse IgG (whole molecule)-Alkaline Phosphatase
F0257FITCAnti-Mouse IgG (whole molecule)-FITC
B7264BiotinAnti-Mouse IgG (whole molecule)-Biotin





Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?