SAB4100029 Sigma

Monoclonal Anti-GBE1 antibody produced in mouse

culture supernatant

DOWNLOAD MSDS (PDF)

Synonym: Anti-Amylo-(1,4 to 1,6) transglucosidase, Anti-Amylo-(1,4 to 1,6) transglycosylase, Anti-GBE, Anti-Glucan (1,4-α-), branching enzyme 1 (GBE1), Anti-Glucan (1,4-α-), branching enzyme 2, Anti-Glycogen branching enzyme, Anti-Glycogen storage disease type IV

Properties

Related Categories Alphabetical Index, Antibodies, Clone Index, G, G-GC,
species reactivity   human
application(s)   western blot: suitable
clone   GBE1-C27, monoclonal
antibody form   culture supernatant
form   aqueous solution
isotype   IgG1
mol wt   antigen mol wt 80.0 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... GBE1(2632)

Description

Immunogen

GBE1 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Price and Availability

Anti-Mouse 2° Antibody Best Sellers


Prod. No.ConjugateSecondary Antibody Description
A4416HRPAnti-Mouse IgG (whole molecule)-Peroxidase
A3562APAnti-Mouse IgG (whole molecule)-Alkaline Phosphatase
F0257FITCAnti-Mouse IgG (whole molecule)-FITC
B7264BiotinAnti-Mouse IgG (whole molecule)-Biotin





Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?