| Related Categories | Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Antibodies to Phosphoproteins at Phosphotyrosine (pTyr), |
| species reactivity | mouse, rat, human |
| application(s) | immunohistochemistry: suitable |
| indirect ELISA: suitable | |
| clone | polyclonal |
| concentration | ~1 mg/mL |
| antibody form | affinity isolated antibody |
| form | buffered aqueous solution |
| mol wt | antigen mol wt 87 kDa |
| shipped in | wet ice |
| storage temp. | −20°C |
| Gene Information | human ... NTRK1(4914) |
| NCBI accession no. |
P04629 |
Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?