Synonym: Anti-Histone H3 (di methyl K4)
| Related Categories | Alphabetical Index, Antibodies, ChIP Validated Antibodies, Epigenetics, H1-HE, |
| species reactivity | human |
| application(s) | ChIP: suitable |
| indirect ELISA: suitable | |
| western blot: suitable | |
| clone | polyclonal |
| concentration | ~1.2 mg/mL |
| antibody form | affinity isolated antibody |
| form | buffered aqueous solution |
| mol wt | antigen mol wt ~17 kDa |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... H3F3A(3021), H3F3B(3021) |
| NCBI accession no. |
NM_002107 |
|
NM_005324 |
Supplied at 1.2 mg/mL in PBS, 0.05% sodium azide., 0.05% ProClin 300.
Imprint is a registered trademark of Sigma-Aldrich Co. LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?