SIHK0095 Sigma

MISSION® siRNA

Human Kinase AKT1 (siRNA2), Nano Scale 0.25 nmol

DOWNLOAD MSDS (PDF)

Properties

Related Categories AA-CG, Functional Genomics and RNAi, MISSION siRNA, siRNA Kinase Nano Scale
storage condition   desiccated
NCBI accession no.   NM_001014431
storage temp.   −20°C
Gene Information   human ... AKT1(207)

Description

General description

The MISSION small scale siRNA are available at 0.25 nmol scale. Sigma′s siRNAs are designed with the Rosetta algorithm and are backed by Sigma-Aldrich’s predesigned siRNA guarantee. You can find all of these off-the-shelf siRNAs on YFG.

Other Notes

Associated siRNA: SASI_Hs01_00205546

Physical form

lyophilized powder

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?