SRP4905 Sigma

Hop human

recombinant, expressed in E. coli, ≥99% (SDS-PAGE)

DOWNLOAD MSDS (PDF)

Synonym: HSC70/HSP90-organizing Protein, Renal carcinoma antigen NY-REN-11, STI1, Stress-induced-phosphoprotein 1, Transformation-sensitive protein IEF SSP 3521

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cell Stress, Chaperonins and Heat Shock Proteins, Heat Shock and Related Proteins,
recombinant   expressed in E. coli
assay   ≥99% (SDS-PAGE)
form   liquid
NCBI accession no.   P31948
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... STIP1(10963)

Description

General description

Hop is HSC70/HSP90-organizing Protein. It is also calledas Stress-induced Phosphoprotein 1, STI1.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.

Physical form

4.5 mg/ml in 40 mM Hepes, pH 7.5.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?