MDL number MFCD00162630
Popular Documents: Datasheet (PDF)
| Related Categories | Alphabetical Index, Antibodies, MO-MR, Mouse Secondary Antibodies and Conjugates, Primary Antibodies, |
| antibody form | affinity isolated antibody |
| clone | polyclonal |
| contains | 15 mM sodium azide |
| storage condition | protect from light |
| application(s) | direct immunofluorescence: 1:32 |
| immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200 | |
| storage temp. | −20°C |
Antibody adsorbed with human IgG.
Adsorbed to reduce background human samples.
Binds mouse IgG; does not bind other mouse Igs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?