| Related Categories | Eppendorf, Eppendorf Pipettors, Eppendorf Pipettors & Tips, Labware, Liquid Handling Equipment, |
| material | yellow control button |
| description | use 100 μl pipette tips. |
| feature | fully autoclavable |
| mfr. no. | Eppendorf, 4900 000.125 (European) |
| Eppendorf, 4900 000.125 | |
| availability | not available in USA and Canada |
| measuring range | 50 μL volume |
Ejector sleeve removable, for easy access to narrow vessels.
Single button control of filling, blow-out and tip ejection.
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG