Z375063 Sigma
Vertical electrophoresis unit, adjustable height, single gel
gel W × H 16.5 cm × 14.5-28 cm, CE-approved
DOWNLOAD MSDS (PDF)
Description
General description
• Made from high quality acrylic with platinum electrodes
• Long lasting silicone sponge gasket on upper reservoir for leak-free operation without grease
• Safety covers with attached safety-tip power leads
• Designed for use with unique gel-casting gasket; heavy-duty casting clamps compress gasket between glass plates, sealing without grease
For Slab Gel Electrophoresis:
• Designed to use as little as 200 mL of buffer, and as little as 5 μg of sample per well
• Use with gel casting accessories which offer the ability to cast one or two gels in a single assembly
• Use in standard slab gel electrophoresis or in the second dimension of 2D electrophoresis
Each unit comes with base unit, upper and lower reservoirs with safety covers and attached power leads, and two gel-mounting spring clamps.
Slab and tube gel accessories are not included; order from the accessories list
Price and Availability
Customers Also Viewed
Vertical electrophoresis unit, adjustable height, dual gel
gel W × H 16.5 cm × 14.5-28 cm, CE-approved
Vertical electrophoresis unit, adjustable height, dual gel
gel W × H 16.5 cm × 14.5-28 cm, Not CE-approved
Vertical electrophoresis unit, adjustable height, single gel
gel W × H 16.5 cm × 14.5-28 cm, Not CE-approved
Vertical electrophoresis unit, fixed height, single gel
Mini, W × H 11.3 cm × 10 cm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?