Z382264 Sigma
Whatman® UNIFILTER® plates
working volume 350 μL, with Whatman GF/B membrane
DOWNLOAD MSDS (PDF)
Properties
Related Categories 96 Well Plates, Core Bioreagents, Labware, Life Science Reagents for Nucleic Acid Purification, Manufacturer Browser,
material   white polystyrene plate
  white polystyrene wells conical bottom
sterility   non-sterile
mfr. no.   Whatman Article No., 28420819 (US reference)
  Whatman, 7700-3303
maximum volume   350 μL
size   96 wells
working volume   350 μL
Description
Application
The UNIFILTER products are convenient, ready to use, disposable filter bottom microplates. Available in 96 well formats, the UNIFILTER microplates offer a choice of filter media that work well with the most stringent applications. UNIFILTERS can be used to process 96 samples at a time. The unique drip director design allows for analysis of the filtrate or the retentate. UNIFILTERS are available in many different variations of well formats and range from 100 μL to 2 mL, using a wide variety of filtration media.
General description
• Design prevents well to well crosstalk
• Large variety of well volumes economizes on expensive consumables
• Choice of filter media allows control of flow rates and retention characteristics
• Standard overall dimensions fit most plate readers, auto samplers, robots
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
UNIFILTER is a registered trademark of GE Healthcare
Whatman is a registered trademark of GE Healthcare
Price and Availability
Customers Also Viewed
Whatman<SUP>®</SUP> UNIFILTER<SUP>®</SUP> plates
working volume 800 μL, with Whatman GF/B membrane
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?