| sterility | sterile |
| feature | ATP free |
| DNA free (Bacterial (E. Coli)) | |
| DNA free (Human) | |
| DNase free | |
| PCR inhibitor free | |
| RNase free | |
| pyrogen free |
Snaps securely into Multipette pipette. Tips are not autoclavable. Biopur tips are certified sterile and pyrogen, RNase-, DNA and ATP-free.
Biopur is a registered trademark of Eppendorf AG
Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG