| Related Categories | Jenco Microscopes, Jenco Stereo Microscopes, Labware, Microscopy |
The GL Series is par-focal and its continuous zoom is available with detentes at 0.5X increments for precise measurements. With superb resolution the GL Series is ideal for semi-conductor, high-tech industries, medical device assembly and life science research.
The comfort design allows for long duration tasks making them less fatiguing with accessible controls. Bonders and probers provide a compact design, and a long working distance, and the microscopes are fully integrated with camera and software systems.
All models include: a pair of 10X eye pieces, black/white reversible stage plate, dust cover, power cord and instruction manual.
• Wide interpupillary adjustment (45 - 75mm) and diopters built-in to the eyepieces
• Side mounted zoom controls and low positioned focus control with tension adjustment
• The clear, non-distorted 3D image resolved by the GL Series optics is a result of advanced technologies in optical design.
• A 100 mm working distance
• Large zoom range gives accurate reproduction of the sample up to 260X
Jenco Boom Stands feature heavy bases for stability and tilting/swiveling mounts to facilitate observation of even hard to get to areas:
Post Stand has a slim 1.25 in. (35 mm) H, streamlined base. Heavy duty design and freedom of movement in multiple directions. The rugged structure minimizes vibration/tipping even with camera attached to trinocular head.
Smooth Boom got its name from the smooth gliding motion produced by the ball bearings in the horizontal movement. Ideal for scanning large parts, like a circuit board, simply with the touch of a hand.
Standard Boom is economical and well-matched to applications where continuous horizontal movement is not required. Used in medical device assembly where work stations and repetitive actions are common.
Articulating Arm Boom features a counter balanced arm that moves vertically as well as horizontally, making it ideal for tight spaces.
Jenco is a trademark of Jenco International, Inc.