| mfr. no. | 4601020 |
| measuring range | , accuracy: 0.07 μL |
| volume | 5 μL |
Uses Finntip 10, 20 or 50 pipet tips
The Finnpipette Focus Fixed Volume pipettes offer the same ergonomic advantages as the rest of the Finnpipette Focus range, including soft-touch tip ejection and the short precision control tip. Finnpipette Fixed Volume pipettes are provided with blue medium sized handles. The entire pipette including the tip cone and snap-on handle can be autoclaved in one piece
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries