| mfr. no. | 4601060 |
| measuring range | , accuracy: 0.3 μL |
| volume | 50 μL |
Uses Finntip 250 Univ, 300 or 200 Ext pipet tips
The Finnpipette Focus Fixed Volume pipettes offer the same ergonomic advantages as the rest of the Finnpipette Focus range, including soft-touch tip ejection and the short precision control tip. Finnpipette Fixed Volume pipettes are provided with blue medium sized handles. The entire pipette including the tip cone and snap-on handle can be autoclaved in one piece
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries