Z708798 Sigma
Harris Uni-Core
I.D. 0.75 mm
DOWNLOAD MSDS (PDF)
Properties
Related Categories Electrophoresis Supplies, Gel Handling, Harris, Labware, Manufacturer Browser,
mfr. no.   Whatman Article No., 29002513 (US reference)
I.D.   0.75 mm
Description
General description
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction, collection and storage in one operation. Suitably samples thin and soft substrates including filter paper, gels and dried blood on cards. Units can be autoclaved at 250°C for limited re-use.
Price and Availability
Customers Also Viewed
Harris cutting mat
L × W × D 8 in. × 6 in. × 1.5 mm
Harris micro-punch
complete, I.D. 1 mm, plastic
Harris Uni-Core
I.D. 0.5 mm
Harris Uni-Core
I.D. 1 mm
Harris Uni-Core
I.D. 1.2 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?