CAS Number 97396-56-0
MDL number MFCD00145830
| Related Categories | Adsorbent Media (Polymeric), Amberlite & Amberchrom, Analytical/Chromatography, Resins & LC Separation Media, Resins and Media More... |
| mfr. no. | Rohm and Haas Amberlite® XAD1180 |
| packaging | pkg of 100 g |
| matrix | styrene/divinylbenzene polyaromatic |
| diameter | 20-60 mesh |
| pore size | 1.68 mL/g pore volume |
| 300 Å pore size | |
| surface area | 600 m2/g |
Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts.
Amberlite® XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water.
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Customers Also Viewed
20-60 mesh
matrix styrene/
20-60 mesh
20-60 mesh
20-60 mesh
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?