24954 Supelco
Fused Silica-Lined Stainless Steel Tubing
tubing L × O.D. × I.D. 6 ft × 1/16 in. (1.59 mm) × 0.02 in. (0.508 mm)
DOWNLOAD MSDS (PDF)
Properties
Related Categories Analytical/Chromatography, Fused Silica-Lineed Stainless Steel Tubing, Gas Chromatography (GC), Gas Purification and Gas Management, Plumbing/Regulation,
tubing L × O.D. × I.D.   6 ft × 1/16 in. (1.59 mm) × 0.02 in. (0.508 mm)
Description
General description
The strength of stainless steel with the inertness of deactivated fused silica. Use for gas transfer lines.
Price and Availability
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?