CAS Number 95-57-8
| Related Categories | Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, C, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 5000 μg/mL in methanol |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?