442629 Supelco

Isopropylamine

anhydrous, analytical standard, for environmental analysis

DOWNLOAD MSDS (PDF)

Synonym: 2-Aminopropane

  • CAS Number 75-31-0

  • Linear Formula (CH3)2CHNH2

  • Molecular Weight 59.11

  •  Beilstein Registry Number 605259

  •  EC Number 200-860-9

  •  MDL number MFCD00008082

  •  PubChem Substance ID 24867934

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard, for environmental analysis
vapor density   2.04 (vs air)
vapor pressure   9.2 psi ( 20 °C)
autoignition temp.   755 °F
quality   anhydrous
expl. lim.   10.4 %
packaging   ampule of 1000 mg
refractive index   n20/D 1.374(lit.)
bp   33-34 °C(lit.)
density   0.688 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Isopropylamine

≥99.5%

Isopropylamine

anhydrous, analytical standard, for environmental analysis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?