442640 Supelco

Methacrylonitrile

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 126-98-7

  • Linear Formula CH2=C(CH3)CN

  • Molecular Weight 67.09

  •  Beilstein Registry Number 773708

  •  EC Number 204-817-5

  •  MDL number MFCD00001872

  •  PubChem Substance ID 24867938

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
vapor pressure   64 mmHg ( 20 °C)
packaging   ampule of 1000 mg
refractive index   n20/D 1.400(lit.)
bp   90-92 °C(lit.)
mp   −35.8 °C(lit.)
density   0.8 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Acrylonitrile

analytical standard, for environmental analysis

Acrylonitrile

≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor

Methacrylic acid

contains 250 ppm MEHQ as inhibitor, 99%

Methacrylonitrile-d<SUB>5</SUB>

97 atom % D, 97% (CP)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?