CAS Number 75-96-7
Linear Formula CBr3CO2H
Molecular Weight 296.74
Beilstein Registry Number 1755150
EC Number 200-919-9
MDL number MFCD00004167
PubChem Substance ID 24867683
| Related Categories | Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Bromo, |
| grade | analytical standard |
| packaging | ampule of 100 mg |
| bp | 245 °C(lit.) |
| mp | 128-132 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?