Synonym:
Acrolein-
CAS Number 888-54-0
Empirical Formula (Hill Notation) C9H8N4O4
Molecular Weight 236.18
EC Number 200-835-2
| Related Categories | A, AA to AL, Air Monitoring, Air Monitoring Standards, Aldehyde/DNPH, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| feature | standard type calibration |
| packaging | ampule of 1 mL |
| concentration | 1000 μg/mL in acetonitrile (as aldehyde equivalent) |
| storage temp. | 2-8°C |
Customers Also Viewed
analytical standard
100 μg/mL in acetonitrile (as aldehyde equivalent), analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?