48238 Supelco

EPA 8040B Phenols Mix

2000 μg/mL each component in isopropanol, analytical standard

DOWNLOAD MSDS (PDF)

Properties

Related Categories 8000 Series Solidwaste Methods, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in isopropanol
suitability   suitable for 8040 per US EPA
storage temp.   2-8°C

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Analyte

Description

Product #

Add to Cart

2-Chlorophenol    
p-Cresol    
2,6-Dichlorophenol    
2,4-Dimethylphenol    
2,4-Dinitrophenol    
Dinoseb    
o-Cresol    
2,3,4,6-Tetrachlorophenol    
2,4,5-Trichlorophenol    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?