Synonym: Heptachlor exo-epoxide solution
CAS Number 1024-57-3
MDL number MFCD01311793
PubChem Substance ID 24872207
| Related Categories | 2000/60/EC, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 20 μg/mL in methanol |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?