48727 Supelco

Benzidine-DFTPP Standard

in methylene chloride (varied), analytical standard

DOWNLOAD MSDS (PDF)

Properties

Related Categories 600 Series Wastewater Methods, A-B, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type tuning solution
packaging   ampule of 1 mL
concentration   in methylene chloride (varied)
suitability   suitable for 625 per US EPA
storage temp.   2-8°C

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Analyte

Description

Product #

Add to Cart

Benzidine 500 μg/mL    
Decafluorotriphenylphosphine 250 μg/mL    

Customers Also Viewed

Decafluorotriphenylphosphine

analytical standard

1000 μg/mL in acetone, analytical standard

250 μg/mL each component in methylene chloride, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?