CAS Number 72-20-8
MDL number MFCD00151158
PubChem Substance ID 24872351
| Related Categories | 2000/60/EC, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 10 mL |
| concentration | 200 μg/mL in isooctane |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?