| Related Categories | Acylation Reagents, Analytical Reagents, Analytical/Chromatography, Derivatization Reagent Sampler Kits, Derivatization Reagents, |
| storage temp. | 2-8°C |
|
Description |
Product # |
Add to Cart |
|---|---|---|
| BF3-Methanol 3 x 1mL | ||
| Methanolic Base 3 x 1mL | ||
| Methanolic HCl (0.5N) 3 x 1mL | ||
| Methanolic HCl (3N) 3 x 1mL | ||
| TMAH, 0.2M in Methanol 3 x 1mL |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?