| composition | bed wt., 50 mg/well |
| packaging | pk of 1 |
Retention Mechanism: Normal phase
Sample Matrix Compatibility: Organic solvents, oils, and lipids
• Polymerically bonded, 2,3-Dihydroxypropoxypropy
• Polar sorbent most commonly used for normal phase applications (polar extractions from non-polar matrices)
• The sorbent′s dihydroxy groups facilitates strong hydrogen bonding
• Excellent selectivity when extracting structurally similar molecules
Discovery is a registered trademark of Sigma-Aldrich Co. LLC