861213 Supelco

EPA Acid Calibration Mix 1

2000 μg/mL each component in methylene chloride, analytical standard

DOWNLOAD MSDS (PDF)

Properties

Related Categories 8000 Series Solidwaste Methods, A, AA to AL, Alphabetical Index of Analytical Standards, Analytical Standards,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 2 mL
concentration   2000 μg/mL each component in methylene chloride
suitability   suitable for 8270 per US EPA

Price and Availability

Analyte

Description

Product #

Add to Cart

2,4-Dinitrophenol    
2-Methyl-4,6-dinitrophenol    
4-Nitrophenol    
Pentachlorophenol    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?