Synonym:
1,1,2-
CAS Number 76-13-1
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Environmental Standards, |
| grade | analytical standard |
| description | Separate Source |
| CofA | certificate of analysis is enclosed in each package. |
| feature | standard type calibration |
| packaging | ampule of 1 × 1 mL (47944) |
| ampule of 1 × 1 mL (4S7944) | |
| concentration | 2000 μg/mL in methanol |
Freon is a registered trademark of E. I. du Pont de Nemours and Company
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?