CAS Number 19666-30-9
Empirical Formula (Hill Notation) C15H18Cl2N2O3
Molecular Weight 345.22
Beilstein Registry Number 558070
MDL number MFCD00128056
PubChem Substance ID 24899098
| grade | analytical standard |
| packaging | ampule of 250 mg |
| mfr. no. | Chem Service Inc, PS-1050 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?