CAS Number 91465-08-6
Empirical Formula (Hill Notation) C23H19ClF3NO3
Molecular Weight 449.85
EC Number 415-130-7
MDL number MFCD07370148
| Related Categories | Acaricides, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, C, |
| grade | analytical standard |
| packaging | ampule of 100 mg |
| mfr. no. | Chem Service Inc, PS-2018 |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?